Merck
  • A multiparametric serum marker panel as a complementary test to mammography for the diagnosis of node-negative early-stage breast cancer and DCIS in young women.
A multiparametric serum marker panel as a complementary test to mammography for the diagnosis of node-negative early-stage breast cancer and DCIS in young women.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology (2014-06-25)
Jérôme Lacombe, Alain Mangé, Anne-Claire Bougnoux, Ioannis Prassas, Jérôme Solassol
ABSTRAKT
The sensitivity of mammography for the detection of small lesions, including node-negative early-stage (T1N0) primary breast cancer (PBC) and ductal carcinoma in situ (DCIS), is significantly decreased in young patients. From a clinical standpoint, an inconclusive mammogram reflects the inability of clinicians to confidently decide whether patients should be referred for biopsy or for follow-up with repeat imaging. Specific ELISAs were developed for a panel of 13 well-recognized breast autoantigens (HSP60, FKBP52, PRDX2, PPIA, MUC1, GAL3, PAK2, P53, CCNB1, PHB2, RACK1, RUVBL1, and HER2). Circulating autoantibody levels were measured in a cohort of 396 serum samples from histologically confirmed DCIS (n = 87) or T1N0 PBC (n = 153) and healthy controls (n = 156). Individually, antibodies against CCNB1, FKBP52, GAL3, PAK2, PRDX2, PPIA, P53, and MUC1 demonstrated discriminatory power between breast cancer and healthy control groups. At 90% sensitivity, the overall combined specificity of the autoantibody serum screening test was 42%. Adjustment for higher sensitivities of 95% and 99% resulted in 30% and 21% specificities, respectively (33% and 18% in T1N0 PBC and 28% and 21% in DCIS). Finally, in patients with node-negative early-stage breast cancer younger than 50 years, the autoantibody assay exhibited 59% specificity with a fixed sensitivity at 90%. Our autoantibody panel allows accurate detection of early breast cancer and DCIS, notably in younger patients. Clinical assessment of this autoantibody panel displays a potential to facilitate clinical management of early-stage breast cancer detection in cases of inconclusive mammogram.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O