Merck
  • Exogenous glucagon-like peptide-2 improves outcomes of intestinal adaptation in a distal-intestinal resection neonatal piglet model of short bowel syndrome.
Exogenous glucagon-like peptide-2 improves outcomes of intestinal adaptation in a distal-intestinal resection neonatal piglet model of short bowel syndrome.
Pediatric research (2014-07-06)
Megha Suri, Justine M Turner, David L Sigalet, Pamela R Wizzard, Patrick N Nation, Ron O Ball, Paul B Pencharz, Patricia L Brubaker, Paul W Wales
ABSTRAKT
Endogenous glucagon-like peptide-2 (GLP-2) levels and intestinal adaptation are reduced in distal-intestinal resection animal models of short bowel syndrome (SBS) that lack remnant ileum. We hypothesized that exogenous GLP-2 would improve intestinal adaptation in a distal-intestinal resection neonatal piglet model of SBS. In all, 35 piglets were randomized to 2 treatment and 3 surgical groups: control (sham), 75% mid-intestinal resection (JI), and 75% distal-intestinal resection (JC). Parenteral nutrition (PN) commenced on day 1 and was weaned as enteral nutrition (EN) advanced. IV GLP-2 (11 nmol/kg/d) or saline was initiated on day 2. Piglets were maintained for 14 d. Clinical, functional, morphological, and histological outcomes were obtained. JC-GLP-2 piglets had fewer days on PN (10.0 ± 0.6 vs. 13.8 ± 0.2), more days on EN (4.0 ± 0.6 vs. 0.2 ± 0.2), a higher percentage of EN at termination (92 ± 5 vs. 52 ± 10%), fewer days of diarrhea (8.0 ± 0.7 vs. 12.3 ± 0.4), increased intestinal length (19 ± 4 vs. -5 ± 3%), and deeper jejunal crypts (248 ± 21 vs. 172 ± 12 μm), compared with saline piglets. GLP-2 therapy improves clinical, morphological, and histological outcomes of intestinal adaptation in a distal-intestinal resection model of SBS. Since this anatomical subtype represents the majority of clinical cases of neonatal SBS, these results support a potential role for GLP-2 therapy in pediatric SBS.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)