Merck
  • Melatonin compensates silencing of heat shock protein 70 and suppresses ultraviolet radiation-induced inflammation in human skin ex vivo and cultured keratinocytes.
Melatonin compensates silencing of heat shock protein 70 and suppresses ultraviolet radiation-induced inflammation in human skin ex vivo and cultured keratinocytes.
Journal of pineal research (2014-11-27)
Konrad Kleszczyński, Stephanie Zwicker, Stefan Tukaj, Michael Kasperkiewicz, Detlef Zillikens, Ronald Wolf, Tobias W Fischer
ABSTRAKT
Melatonin, a lipophilic compound synthesized and released from the pineal gland, effectively acts against ultraviolet radiation (UVR), one of the main inducers of epidermal damage, skin cancer, inflammation, and DNA photo damage. One of the common known stress protein induced by UVR is heat shock protein 70 (Hsp70), highly expressed in human keratinocytes, providing cellular resistance to such stressors. Here, using human full-thickness skin and normal human epidermal keratinocytes (NHEK), we investigated the interaction of melatonin and Hsp70 toward UVR-induced inflammatory and apoptotic responses. The following observations were made: (i) UVR upregulated Hsp70 gene expression in human epidermis while melatonin significantly inverted this effect, (ii) similar patterns of regulation were observed within Hsp70 protein level, and (iii) mechanistic studies involving silencing of Hsp70 RNA (Hsp70 siRNA) showed prominent decrease of IκB-α (an inhibitor of NF-κB) and enhanced gene expression of pro-inflammatory cytokines (IL-1β, IL-6, Casp-1) and pro-apoptotic protein (Casp-3) in NHEK. Parallel investigation using melatonin (10(-3)  m) significantly inverted these responses regardless depletion of Hsp70 RNA suggesting a compensatory action of this compound in the defense mechanisms. Our findings combined with data reported so far thus enrich existing knowledge about the potent anti-apoptotic and anti-inflammatory action of melatonin.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
3,3′,5,5′-Tetramethylbenzidine, standard for GC
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Acetone, natural, ≥97%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Sigma-Aldrich
Melatonin, powder, ≥98% (TLC)
SAFC
L-Glutamine
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
USP
Melatonin, United States Pharmacopeia (USP) Reference Standard