Merck
  • Evaluation of various silicon-and boron-containing compounds for the detection of phosphorylation in peptides via gas-phase ion-molecule reactions.
Evaluation of various silicon-and boron-containing compounds for the detection of phosphorylation in peptides via gas-phase ion-molecule reactions.
European journal of mass spectrometry (Chichester, England) (2014-11-26)
Andrii Piatkivskyi, Yuriy Pyatkivskyy, Victor Ryzhov
ABSTRAKT
Gas-phase ion-molecule reactions [IMR] of various boron- and silicon-containing neutrals were investigated as a potential route for detecting phosphorylation within peptides in the negative ion mode. Trimethyl borate (TMB), triethyl borate (TEB) and N,O- Bis(trimethylsilyl)acetamide (TMSA), unlike diethylmethoxyborane (DEMB), diisopropoxymethylborane [DiPMB] and chlorotrimethylsi- Lane (TMSCIL], reacted differently if a phosphate moiety was present and thus are suitable to detect phosphorylation. During multistage collision-induced dissociation experiments of the reaction products of IMR with TMB and TEB, the [LSsF - 4H + B]- ion formed a modified y2 fragment allowing the phosphorylation site to be assigned, unlike reaction products of DEMB and DiPMB which lost both the phos- phoric acid and the boron-containing moiety.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard
Supelco
Chlorotrimethylsilane, derivatization grade (GC derivatization), LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, Wacker Chemie AG, ≥99.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, puriss., ≥99.0% (GC)
Serine, European Pharmacopoeia (EP) Reference Standard
Leucine, European Pharmacopoeia (EP) Reference Standard
Methionine, European Pharmacopoeia (EP) Reference Standard
DL-Methionine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DL-Methionine, 99%, FCC, FG
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
DL-Leucine, ≥99% (HPLC)
Sigma-Aldrich
Chlorotrimethylsilane solution, 1.0 M in THF
Sigma-Aldrich
DL-Methionine, ≥99.0% (NT)
Sigma-Aldrich
DL-Methionine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
DL-Methionine, ≥99%
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)