Merck
  • Can biomarkers differentiate pain and no pain subgroups of nonverbal children with cerebral palsy? A preliminary investigation based on noninvasive saliva sampling.
Can biomarkers differentiate pain and no pain subgroups of nonverbal children with cerebral palsy? A preliminary investigation based on noninvasive saliva sampling.
Pain medicine (Malden, Mass.) (2014-09-23)
Frank J Symons, Issam ElGhazi, Brian G Reilly, Chantel C Barney, Leah Hanson, Angela Panoskaltsis-Mortari, Ian M Armitage, George L Wilcox
ABSTRAKT
Assessing and treating pain in nonverbal children with developmental disabilities are a clinical challenge. Current assessment approaches rely on clinical impression and behavioral rating scales completed by proxy report. Given the growing health relevance of the salivary metabolome, we undertook a translational-oriented feasibility study using proton nuclear magnetic resonance (NMR) spectroscopy and neuropeptide/cytokine/hormone detection to compare a set of salivary biomarkers relevant to nociception. Within-group observational design. Tertiary pediatric rehabilitation hospital. Ten nonverbal pediatric patients with cerebral palsy with and without pain. Unstimulated (passively collected) saliva was collected using oral swabs followed by perchloric acid extraction and analyzed on a Bruker Avance 700 MHz NMR spectrometer. We also measured salivary levels of several cytokines, chemokines, hormones, and neuropeptides. Partial least squares discriminant analysis showed separation of those children with/without pain for a number of different biomarkers. The majority of the salivary metabolite, neuropeptide, cytokine, and hormone levels were higher in children with pain vs no pain. The ease of collection and noninvasive manner in which the samples were collected and analyzed support the possibility of the regular predictive use of this novel biomarker-monitoring method in clinical practice.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Perchloric acid, 0.01 M HClO4 in water (0.01N), eluent for IC
Sigma-Aldrich
Perchloric acid, 70%, 99.999% trace metals basis
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, 70 atom % D
Sigma-Aldrich
Deuterium oxide, 60 atom % D
Sigma-Aldrich
Potassium hydroxide, tested according to Ph. Eur.
Sigma-Aldrich
Potassium hydroxide, ≥99.95% trace metals basis
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Deuterium oxide, 99.8 atom % D
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Potassium hydroxide, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Perchloric acid, ACS reagent, 60%
Sigma-Aldrich
Perchloric acid, ACS reagent, 70%
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard