Merck
Administered chrysanthemum flower oil attenuates hyperuricemia: mechanism of action as revealed by DNA microarray analysis.
Bioscience, biotechnology, and biochemistry (2014-07-19)
Shinichi Honda, Seiji Kawamoto, Hozumi Tanaka, Hideyuki Kishida, Masayasu Kitagawa, Yuji Nakai, Keiko Abe, Dai Hirata
ABSTRAKT
We applied Chrysanthemum flower oil (CFO) to a hyperuricemia model by feeding rats a hyperuricemia-inducing diet (HID) and investigated its effect on serum uric acid (SUA) levels and its mode of action. CFO is the oily fraction that contains polyphenols derived from chrysanthemum flowers. Oral administration of CFO to HID-fed rats significantly decreased their SUA levels. It also inhibited xanthine oxidase activities in the liver and increased urine uric acid levels. The effects of CFO on the renal gene expressions that accompanied the induction of hyperuricemia were comprehensively confirmed by DNA microarray analysis. The analysis showed up-regulation of those genes for uric acid excretion by CFO administration. These results suggest that CFO suppresses the increase in SUA levels via two mechanisms: suppression of uric acid production by inhibition of xanthine oxidase in the liver and acceleration of its excretion by up-regulation of uric acid transporter genes in the kidney.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Uric acid, ≥99%, crystalline
Sigma-Aldrich
Uric acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Oxonic acid potassium salt, 97%
Sigma-Aldrich
Xanthine, ≥99%
Sigma-Aldrich
Xanthine, ≥99.5% (HPLC), purified by recrystallization
Sigma-Aldrich
Xanthine, BioUltra, ≥99%