Merck
The effect of HPMC particle size on the drug release rate and the percolation threshold in extended-release mini-tablets.
Drug development and industrial pharmacy (2013-10-19)
Faiezah A A Mohamed, Matthew Roberts, Linda Seton, James L Ford, Marina Levina, Ali R Rajabi-Siahboomi
ABSTRAKT
The particle size of HPMC is a critical factor that can influence drug release rate from hydrophilic matrix systems. Percolation theory is a statistical tool which is used to study the disorder of particles in a lattice of a sample. The percolation threshold is the point at which a component is dominant in a cluster resulting in significant changes in drug release rates. Mini-tablets are compact dosage forms of 1.5-4 mm diameter, which have potential benefits in the delivery of drug to some patient groups such as pediatrics. In this study, the effect of HPMC particle size on hydrocortisone release and its associated percolation threshold for mini-tablets and tablets was assessed. For both mini-tablets and tablets, large polymer particles reduced tensile strength, but increased the drug release rate and the percolation threshold. Upon hydration, compacts with 45-125 μm HPMC particles formed a strong gel layer with low porosity, reducing hydrocortisone release rates. In comparison, faster drug release rates were obtained when 125-355 µm HPMC particles were used, due to the greater pore sizes that resulted in the formation of a weaker gel. Using 125-355 µm HPMC particles increased the percolation threshold for tablets and to a greater extent for mini-tablets. This work has demonstrated the importance of HPMC particle size in ER matrices, the effects of which are even more obvious for mini-tablets.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Magnesium stearate, puriss., meets analytical specification of Ph. Eur., BP, ≥90% stearic and palmitic acid basis, ≥40% stearic acid basis (GC), 4.0-5.0% Mg basis (calc on dry sub.)
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, PCR Reagent
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Magnesium stearate, technical grade
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Supelco
Standardowa gęstość 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, for ultratrace analysis, ACS reagent
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, Deionized
Supelco
Water, for TOC analysis
Supelco
Water, suitable for ion chromatography
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Supelco
Water, suitable for HPCE, suitable for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, tested according to Ph. Eur.
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard