Merck
Comparative endothelial profiling of doxorubicin and daunorubicin in cultured endothelial cells.
Toxicology in vitro : an international journal published in association with BIBRA (2014-12-23)
Tomasz Wojcik, Elzbieta Buczek, Katarzyna Majzner, Agnieszka Kolodziejczyk, Justyna Miszczyk, Patrycja Kaczara, Wojciech Kwiatek, Malgorzata Baranska, Marek Szymonski, Stefan Chlopicki
ABSTRAKT
Although anthracycline antibiotics have been successfully used for nearly half a century in the treatment of various malignancies, their use is limited by their cardiac and vascular toxicities, and the mechanisms of these toxicities are still not entirely clear. Herein, we comprehensively characterized cytotoxic effects of two structurally related anthracyclines, doxorubicin and daunorubicin. In nanomolar concentrations, both drugs induced DNA damage and increased nuclear area that were associated with their accumulation in the nucleus (doxorubicin ⩾50 nM and daunorubicin ⩾25 nM) as evidence by Raman microspectroscopy at 3820-4245 cm(-1). At low micromolar concentrations, doxorubicin (⩾5 μM) and daunorubicin (⩾1 μM) increased the generation of reactive oxygen species, decreased intracellular reduced glutathione, induced an alteration in endothelial elasticity and caused a reorganization of the F-actin cytoskeleton. In isolated mouse aortic rings, doxorubicin (⩾50 μM) was less potent than daunorubicin (⩾5 μM) in impairing the endothelium-dependent response. In summary, using a comprehensive endothelial profiling approach, we demonstrated clear-cut differences in the potencies to induce endotheliotoxic responses for two structurally similar chemotherapeutics, at a nuclear, cytosolic and membrane levels. Furthermore, our results suggest that the differences in the endothelial toxicities of doxorubicin and daunorubicin are linked to differences in their nuclear accumulation and the DNA damage-triggered response of the endothelium.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Calcium fluoride, random crystals, 99.99% trace metals basis
Sigma-Aldrich
Daunorubicin hydrochloride, ≥90% (HPLC)
Sigma-Aldrich
Daunorubicin hydrochloride, meets USP testing specifications
Daunorubicin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Calcium fluoride, powder, 99.99% trace metals basis
Doxorubicin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dihydroethidium, BioReagent, suitable for fluorescence, ≥95% (HPCE)
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Dihydroethidium, ≥95%
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
USP
Doxorubicin hydrochloride, United States Pharmacopeia (USP) Reference Standard