Merck
Artesunate induces apoptosis of bladder cancer cells by miR-16 regulation of COX-2 expression.
International journal of molecular sciences (2014-09-10)
Wei Zuo, Zhen-Zhong Wang, Jun Xue
ABSTRAKT
Bladder cancer is the most common malignant tumor of the urinary tract and remains one of the major causes of cancer death worldwide. In this study, we investigated the effect and mechanism of Artesunate (ART), a traditional Chinese medicine, on inducing apoptosis of human bladder cancer cells. In vivo antitumor activity was investigated in bladder cancer in rat by subcutaneous injection of different concentration of ART. The effect of ART on growth inhibition and apoptosis of bladder cancer cells was evaluated using dimethylthiazoly-2,5-diphenyltetrazolium bromide (MTT) assay and flow cytometry analysis, respectively. Cyclooxygenase-2 (COX-2) and miR-16 expression levels were determined with real-time PCR. The concentrations of prostaglandin E2 (PGE2) in the supernatants of bladder cancer cells were measured with an ELISA kit. The miR-16 inhibitor or mimic were transfected into cells to up- or down-regulate miR-16 expression. ART efficiently inhibited orthotopic tumor growth in the bladder cancer rat, which is accompanied with an increase of miR-16 expression and a decrease of COX-2 expression. In vitro, ART could induce cytotoxicity and apoptosis in bladder cancer cells, but presented a much lighter toxicity effect against normal human urothelial cells. ART significantly increased miR-16 expression and decreased the expression of COX-2 and the production of PGE2. More importantly, down-regulation of miR-16 expression could reverse the effect of ART on apoptosis and COX-2 expression in bladder cells. Moreover, exogenous PGE2 could inhibit apoptosis of bladder cancer cells treated with ART. In conclusion, ART can elicit an anti-tumor effect against bladder cancer by up-regulation of miR-16 expression, which resulted in the decrease of COX-2 expression and PGE2 production. Hence, ART might be an effective drug for the treatment of bladder cancer.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Dinoprostone, European Pharmacopoeia (EP) Reference Standard