Merck
  • An observational study examining the relationship between respiratory symptoms, airway inflammation and bacteriology in children with severe neurodisability.
An observational study examining the relationship between respiratory symptoms, airway inflammation and bacteriology in children with severe neurodisability.
PloS one (2015-04-09)
Ruth E Trinick, Lara Bunni, Kent Thorburn, Angela P Hackett, Mark Dalzell, Paul S McNamara
ABSTRAKT
Children with severe neurodisability (ND) commonly suffer from chronic respiratory symptoms that impact greatly on quality of life, and lead to recurrent hospital admissions. This morbidity (and its causes) is poorly described, despite being well recognised by paediatricians. In this study, we characterised respiratory symptoms in children with ND at times of stability and deterioration. We also assessed the relationship between respiratory symptoms, lower airway inflammatory markers and levels of infection/colonisation. ND children were recruited upon admission for elective surgery (Elective-ND [n = 16]), or acutely upon admission to Intensive Care (PICU-ND [n = 19]), and compared to healthy control children [n = 12]. Parents completed a validated respiratory symptom questionnaire in which symptoms associated with activity were removed (total maximal score of 108). Bronchoalveolar lavage (BAL) was collected, and BAL neutrophil counts, IL-8 and TGFβ-1 levels measured. BAL microbial analysis was performed using a 16S/18S rRNA gene based assay and Pseudomonas aeruginosa PCR. All ND children had high levels of respiratory symptoms (median [IQR] symptom score PICU-ND, 55[38-64]; Elective-ND, 26[7-45]; Control, 4[0-7]: p<0.01), which affected their families, particularly at nighttime. Elective-ND patients with a total respiratory symptom score >20 invariably had BAL neutrophilia. Elective patients with 16S/18S microbial rDNA positive BAL had higher neutrophil counts (positive, 33[18-70]%; negative, 8[4-38]%: p<0.05) and generally higher symptom scores (positive, 17[5-32]; negative, 5[0-9]: p = 0.097). Streptococcus mitis was commonly identified in BAL from ND children; Pseudomonas aeruginosa was not identified in any sample. Children with severe ND often have high levels of chronic respiratory symptoms, which may relate to lower airway inflammation. Bacterial airway colonisation, particularly with oral commensals, may play a role in both symptom generation and inflammation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
2,3-Dimercapto-1-propanol, ≥98% (iodometric)
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis