Merck
Epstein-Barr virus infection induces an increase of T regulatory type 1 cells in Hodgkin lymphoma patients.
British journal of haematology (2014-07-22)
Olivier Morales, Dhafer Mrizak, Violaine François, Rami Mustapha, Céline Miroux, Stéphane Depil, Anne-Valérie Decouvelaere, Pauline Lionne-Huyghe, Claude Auriault, Yvan de Launoit, Véronique Pancré, Nadira Delhem
ABSTRAKT
Epstein-Barr Virus (EBV) is present in the neoplastic cells of around 20-30% of patients with Hodgkin Lymphoma (HL). Although, an immunosuppressive environment is currently described in HL patients, little is known concerning the regulatory mechanism induced by EBV proteins expression in tumour cells. This study aimed to investigate an association between regulatory Type 1 cells (Tr1) and EBV tissue positivity in HL patients. Transcriptomic analysis of both EBV-positive and EBV-negative tumours showed that EBV infection increased gene expression of Tr1-related markers (ITGA2, ITGB2, LAG3) and associated-immunosuppressive cytokines (IL10). This up-regulation was associated with an over-expression of several chemokine markers known to attract T-helper type 2 (Th2) and regulatory T cells thus contributing to immune suppression. This Tr1 cells recruitment in EBV-positive HL was confirmed by immunohistochemical analysis of frozen nodes biopsies and by flow cytometric analysis of peripheral blood mononuclear cells of EBV-positive patients. Additionally, we showed that IL10 production was significantly enhanced in tumours and blood of EBV-positive HL patients. Our results propose a new model in which EBV can recruit Tr1 cells to the nodes' microenvironment, suggesting that the expression of EBV proteins in tumour cells could enable the escape of EBV-infected tumour cells from the virus-specific CTL response.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
USP
Alkohol odwodniony, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications