Merck
Quantitation of Maillard reaction products in commercially available pet foods.
Journal of agricultural and food chemistry (2014-08-05)
Charlotte van Rooijen, Guido Bosch, Antonius F B van der Poel, Peter A Wierenga, Lucille Alexander, Wouter H Hendriks
ABSTRAKT
During processing of pet food, the Maillard reaction occurs, which reduces the bioavailability of essential amino acids such as lysine and results in the formation of advanced Maillard reaction products (MRPs). The aim of this study was to quantitate MRPs (fructoselysine (FL), carboxymethyllysine (CML), hydroxymethylfurfural (HMF)) and the cross-link lysinoalanine (LAL) in commercial pet foods. Sixty-seven extruded, canned, and pelleted dog and cat foods for growth and maintenance were analyzed using UPLC-MS. Canned pet foods contained on average the most FL, CML, and HMF (4534, 37, and 1417 mg/kg dry matter, respectively) followed by pelleted and extruded foods. Average daily intake (mg/kg body weight(0.75)) of HMF is 122 times higher for dogs and 38 times higher for cats than average intake for adult humans. As commercial pet foods are most often the only source of food for dogs and cats, future research focus should be on the bioavailability and long-term health implications of MRP consumption by dogs and cats.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Perfluoropentanoic acid, 97%
Sigma-Aldrich
Furfural, natural, ≥98%, FCC, FG
Sigma-Aldrich
Furfural, 99%
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Furfural, ACS reagent, 99%
Supelco
Diethyl ether, analytical standard
Sigma-Aldrich
Furfural, ≥98%, FCC, FG
Sigma-Aldrich
Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethyl ether, suitable for HPLC, ≥99.9%, inhibitor-free
Supelco
Furfural
Sigma-Aldrich
Zirconyl chloride octahydrate, reagent grade, 98%
Sigma-Aldrich
Diethyl ether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethyl ether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor