Merck
Lignan formation in hairy root cultures of Edelweiss (Leontopodium nivale ssp. alpinum (Cass.) Greuter).
Fitoterapia (2014-06-17)
Christoph Wawrosch, Stefan Schwaiger, Hermann Stuppner, Brigitte Kopp
ABSTRAKT
A hairy root line of Edelweiss (Leontopodium nivale ssp. alpinum (Cass.) Greuter) was obtained upon transformation with Agrobacterium rhizogenes strain ATCC15834. Elicitation of this line with silver nitrate, sucrose, methyl jasmonate and yeast extract at various concentrations in most cases resulted in a stimulation of lignan biosynthesis. Through elicitation with 6% sucrose the roots accumulated the pharmacologically active lignans leoligin and 5-methoxy-leoligin at levels of 0.0678% and 0.0372%, respectively, without significant growth inhibition. These lignan levels were comparable to those found in intact roots of cultivated Edelweiss. The biotechnological production of leoligin could be an attractive option for the continuous, field culture-independent production of the valuable secondary metabolites leoligin and 5-methoxy-leoligin.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sucrose, European Pharmacopoeia (EP) Reference Standard
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
USP
Rozpuszczalnik resztkowy klasy 2 - acetonitryl, United States Pharmacopeia (USP) Reference Standard
Supelco
Dichloromethane, analytical standard