Merck
  • Single course of antenatal betamethasone produces delayed changes in morphology and calbindin-D28k expression in a rat's cerebellar Purkinje cells.
Single course of antenatal betamethasone produces delayed changes in morphology and calbindin-D28k expression in a rat's cerebellar Purkinje cells.
Acta neurobiologiae experimentalis (2015-01-13)
Rodrigo Pascual, Martina Valencia, Sebastián Larrea, Carlos Bustamante
ABSTRAKT
In the current study, we analyzed the impact of antenatal betamethasone on macroscopic cerebellar development, Purkinje cell morphology and the expression of the neuroprotective protein calbindin-D28k. Pregnant rats (Sprague-Dawley) were randomly divided into two experimental groups: control (CONT) and betamethasone-treated (BET). At gestational day 20 (G20), BET dams were subcutaneously injected with a solution of 0.17 mg kg⁻¹ of betamethasone, while CONT animals received a similar volume of saline. At postnatal days 22 (P22) and P52, BET and CONT offspring were behaviorally evaluated, and the cerebella were histologically and immunohistochemically processed. Animals that were prenatally treated with a single course of betamethasone exhibited long-lasting behavioral changes consistent with anxiety-like behavior in the open-field test, together with (1) reduced cerebellar weight and volume, (2) Purkinje cell dendritic atrophy, and (3) an overexpression of calbindin-D28k. The current results indicate that an experimental single course of betamethasone in pregnant rats produces long-lasting anxiety-like behaviors, together with macroscopic and microscopic cerebellar alterations.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Isoflurane, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Supelco
Oxalic acid concentrate, 0.1 M (COOH)2 (0.2N), eluent concentrate for IC
Sigma-Aldrich
Oxalic acid, puriss. p.a., anhydrous, ≥99.0% (RT)
Sigma-Aldrich
Oxalic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Oxalic acid, purified grade, 99.999% trace metals basis
Sigma-Aldrich
Oxalic acid, 98%
Sigma-Aldrich
Zirconyl chloride octahydrate, reagent grade, 98%
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Acetone, natural, ≥97%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sucrose, meets USP testing specifications
Betamethasone, European Pharmacopoeia (EP) Reference Standard
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sucrose, European Pharmacopoeia (EP) Reference Standard
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Betamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Betamethasone, United States Pharmacopeia (USP) Reference Standard
Supelco
Betamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Betamethasone, ≥98%
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra