Merck
Removal of hexavalent chromium from aqueous solution using exfoliated polyaniline/montmorillonite composite.
Water science and technology : a journal of the International Association on Water Pollution Research (2014-08-15)
Jun Chen, Xiaoqin Hong, Yongteng Zhao, Qianfeng Zhang
ABSTRAKT
Exfoliated polyaniline/montmorillonite (PANI/MMT) composites with nanosheet structure were successfully prepared by in situ chemical oxidation polymerization with MMT platelets as the scaffold. Amphoteric polymer, (2-methacryloyloxyethyl)trimethyl ammonium chloride and methacrylate acid copolymer, was used to modify montmorillonite and a large number of carboxylic acids were introduced on the surface of the clay platelets, which can be used as a dopant of PANI and play a 'bridge' role to combine PANI with clay. Adsorption experiments were carried out to study the effects of pH, contact time, Cr(VI) concentration, adsorbent dose and temperature. The adsorption of Cr(VI) on the PANI/MMT was highly pH dependent and the adsorption kinetics followed a pseudo-second-order model. The Langmuir isothermal model described the adsorption isotherm data well and the maximum adsorption capacity increased with the increase in temperature. Thermodynamic investigation indicated that the adsorption process is spontaneous, endothermic and marked with an increase in randomness at the adsorbent - liquid interface. The maximum adsorption capacity of the PANI/MMT composites for Cr(VI) was 308.6 mg/g at 25 °C. The excellent adsorption characteristic of exfoliated PANI/MMT composites will render it a highly efficient and economically viable adsorbent for Cr(VI) removal.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Ammonium persulfate, BioUltra, ≥98.0% (RT)
Sigma-Aldrich
Ammonium persulfate, ACS reagent, ≥98.0%
Sigma-Aldrich
Ammonium persulfate, free-flowing, Redi-Dri, reagent grade, 98%
Sigma-Aldrich
Ammonium persulfate, BioXtra, ≥98.0%
Sigma-Aldrich
Ammonium persulfate, Molecular Biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
Ammonium persulfate, reagent grade, 98%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Aniline, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Aniline, analytical standard
Sigma-Aldrich
Aniline, ReagentPlus®, 99%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O