Merck
Combination of BMP2 and MSCs significantly increases bone formation in the rat arterio-venous loop model.
Tissue engineering. Part A (2014-08-20)
Gregor Buehrer, Amelie Balzer, Isabel Arnold, Justus P Beier, Carolin Koerner, Oliver Bleiziffer, Andreas Brandl, Christian Weis, Raymund E Horch, Ulrich Kneser, Andreas Arkudas
ABSTRAKT
In this study the induction of bone formation in an axially vascularized bone matrix using mesenchymal stem cells (MSCs) and application of bone morphogenetic protein 2 (BMP2) was analyzed in the arteriovenous loop (AVL) model. An AVL was created in the medial thigh of 42 rats and placed in a porous titanium chamber filled with a particulated porous hydroxyapatite and beta-tricalcium phosphate matrix and fibrin. In group A the fibrin was loaded with 5×10(6) DiI-stained fibrin gel-immobilized primary MSCs from syngenic Lewis rats, in group B the matrix was loaded with 60 μg/mL BMP2 and in group C both, BMP2 and MSCs were applied at implantation time point. After 6 and 12 weeks, specimens were investigated by means of histological, morphometrical, and micro-computed tomography analysis. After implantation of an AVL a dense vascular network was visible in all groups. In group A, newly generated bone islands were detected in the periphery of the main vascular axis. Using BMP2 alone (group B), small islands of newly formed bone were visible evenly distributed in all parts of the constructs. In group C nearly the whole matrix was interspersed with bone formations. In all groups there was an increase of bone formation between the 6 and 12 weeks explantation time points. This study demonstrates for the first time the successful generation of axially vascularized bone substitutes using MSCs and BMP2 in the AVL rat model using a one step procedure. Using the combination of BMP2 and MSCs there was a significant increase of bone formations detectable compared to the BMP2 or MSCs alone groups.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), contains 5 wt. % silica as dopant, synthetic
Sigma-Aldrich
Hydroxyapatite, nanoparticles, dispersion, 10 wt. % in H2O, <200 nm particle size (BET)
Sigma-Aldrich
Hydroxyapatite, powder, synthetic
Sigma-Aldrich
Hydroxyapatite, synthetic, 99.8% trace metals basis (excludes Mg)
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
Calcium phosphate tribasic, BioReagent, suitable for plant cell culture, powder
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine perchlorate, BioReagent, suitable for fluorescence, ≥98.0% (TLC)
Sigma-Aldrich
Hydroxyapatite, puriss., meets analytical specification of Ph. Eur., BP, FCC, E341, ≥90% (calculated on glowed substance)
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), ≥97%, synthetic
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine perchlorate, 97%
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Mannitol, European Pharmacopoeia (EP) Reference Standard
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydroxyapatite, purum p.a., ≥90% (as Ca3(PO4)2, KT)
Sigma-Aldrich
Calcium phosphate tribasic, 34.0-40.0% Ca basis
Isoflurane, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
α-Tri-Calcium phosphate, Reagent for transient & stable DNA transfections
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Hematoxylin