Merck
Breast cancer cells imaging by targeting methionine transporters with gadolinium-based nanoprobe.
Molecular imaging and biology : MIB : the official publication of the Academy of Molecular Imaging (2014-02-07)
Bita Mehravi, Mehdi Shafiee Ardestani, Maryam Damercheli, Haleh Soltanghoraee, Negar Ghanaldarlaki, Ali M Alizadeh, Mohammad A Oghabian, Maryam Shahzad Shirazi, Shabnam Mahernia, Massoud Amanlou
ABSTRAKT
Early cancer diagnosis using MRI imaging is of high global interest as a non-invasive and powerful modality. In this study, methionine was conjugated on gadolinium-based mesoporous silica nanospheres to evaluate intra-cellular uptake and its accumulation in human breast cancer cells. The contrast agent was synthesized and characterized using different techniques including N2 physisorption, thermal gravimetric analysis, dynamic light scattering, and inductively coupled plasma atomic emission spectroscopy (ICP-AES). The intra-cellular uptake of Gd(3+) was measured by ICP-AES, fluorescent microscopy, and flow cytometry. Finally, cellular and tumor MR imaging were performed to determine in vitro and in vivo relaxometry. According to the results, the contrast agents accumulated in tumor cells both in vitro and in vivo. There was no significant cellular toxicity on either normal or cancer cells along with strong intense signal on T 1 compared to the unlabeled cells. The results showed that the novel contrast agent could become a useful tool in early detection of cancer.
MATERIAŁY
Numer produktu
Marka
Opis produktu
USP
Bromek cetrymonium, United States Pharmacopeia (USP) Reference Standard
USP
Alkohol odwodniony, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formamide, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Formamide, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Formamide, spectrophotometric grade, ≥99%
Sigma-Aldrich
Diethylenetriamine, ReagentPlus®, 99%
Sigma-Aldrich
Formamide, ≥99.5% (GC), BioReagent, Molecular Biology
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Supelco
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Supelco
Formamide solution, suitable for NMR (reference standard), 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Supelco
Formamide solution, suitable for NMR (reference standard), 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.