Merck
The occurrence of anti-retroviral compounds used for HIV treatment in South African surface water.
Environmental pollution (Barking, Essex : 1987) (2015-02-15)
Timothy Paul Wood, Cornelia S J Duvenage, Egmont Rohwer
ABSTRAKT
The study and quantification of personal care products, such as pharmaceuticals, in surface water has become popular in recent years; yet very little description of these compounds' presence in South African surface water exists in the literature. Antiretrovirals (ARVs), used to treat human immunodeficiency virus (HIV) are rarely considered within this field. A new method for the simultaneous quantification of 12 antiretroviral compounds in surface water using the standard addition method is described. Water samples were concentrated by a generic automated solid phase extraction method and analysed by ultra-high pressure liquid chromatography tandem mass spectrometry (UHPLC-MS/MS). Substantial matrix effect was encountered in the samples with an average method detection limit of 90.4 ng/L. This is the first reported countrywide survey of South African surface water for the quantification of these compounds with average concentrations ranging between 26.5 and 430 ng/L.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Melting point standard 235-237°C, analytical standard
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, analytical standard
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Caffeine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Caffeine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%