Merck
  • Time-kill effect of levofloxacin on multidrug-resistant Pseudomonas aeruginosa and Acinetobacter baumannii: synergism with imipenem and colistin.
Time-kill effect of levofloxacin on multidrug-resistant Pseudomonas aeruginosa and Acinetobacter baumannii: synergism with imipenem and colistin.
European journal of clinical microbiology & infectious diseases : official publication of the European Society of Clinical Microbiology (2014-09-07)
A Safarika, I Galani, A Pistiki, E J Giamarellos-Bourboulis
ABSTRAKT
In the present study, we challenged the concept that levofloxacin should not be used for the management of ventilator-associated pneumonia (VAP) when minimum inhibitory concentrations (MICs) exceed 2 μg/ml. Multidrug-resistant (MDR) and genetically distinct isolates of Pseudomonas aeruginosa (n = 49) and Acinetobacter baumannii (n = 29) from patients with VAP were exposed over time to levofloxacin, imipenem, colistin and their combinations. Synergy between levofloxacin and imipenem was found in 55.3 % and between levofloxacin and colistin in 90.9 % of isolates of P. aeruginosa within the first 4 h of growth. Synergy with imipenem but not with colistin was dependent of the MIC. Synergy between levofloxacin and imipenem was found in 58.6 % of isolates of A. baumannii after 24 h of growth. Considerable synergy was found between levofloxacin and colistin, reaching 84.8 % of isolates of A.baumannii after 6 h of growth. Synergy was independent from the MIC. These results create hopes that levofloxacin can be used as combination therapy for infections by MDR bacteria.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Levofloxacin, analytical standard
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Levofloxacin, 98.0-102.0% anhydrous basis (HPLC)
Informacje o cenach i dostępności nie są obecnie dostępne.
USP
Lewofloksacyna, United States Pharmacopeia (USP) Reference Standard
Informacje o cenach i dostępności nie są obecnie dostępne.