Merck
In-vitro melanoma models: invasive growth is determined by dermal matrix and basement membrane.
Melanoma research (2014-06-04)
Suzan Commandeur, Sarah J Sparks, Hee-Lam Chan, Linda Gao, Jacoba J Out, Nelleke A Gruis, Remco van Doorn, Abdoelwaheb El Ghalbzouri
ABSTRAKT
A critical first step in the metastatic progression of cutaneous melanoma, invasive growth into the dermal compartment, would ideally be studied in the proper three-dimensional tissue microenvironment. In this study, we compared the growth and behavior of four melanoma cell lines originating from primary and metastatic human cutaneous melanomas (AN, RU, M14, and WK) in in-vitro human skin equivalents (HSEs) generated with four different dermal matrices: human fibroblast-seeded rat tail collagen, human fibroblast-derived matrix (FDM), noncellular human de-epidermized dermis (DED), and a novel fully cellular human DED with an intact pre-existent basement membrane. Melanoma cells showed proliferation in all HSEs, indicating that the microenvironment formed in all HSEs studied here allows the growth of melanoma cells in concert with epidermal keratinocytes for multiple weeks in vitro. Melanoma cells did not affect epidermal proliferation and terminal differentiation. Growth of melanoma cells in the dermal compartment, as a measure of invasive potential, differs markedly between the four types of in-vitro human melanoma models. Notably, the growth of melanoma cells in the dermal matrix was observed in all HSEs cultured with cell lines originating from metastatic melanoma, except for cDED-based HSEs, and the growth of melanoma cells of nonmetastatic origin was observed in the dermal compartment of FDM-based HSEs. Our results show that the type of dermal equivalent and the presence of an intact basement membrane should be taken into consideration when studying melanoma invasion using in-vitro HSEs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Palmitic acid, United States Pharmacopeia (USP) Reference Standard
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Palmitic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Supelco
Palmitic acid, analytical standard
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Supelco
Palmitic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Palmitic acid, natural, 98%, FG
Supelco
Linoleic acid, analytical standard