Merck
  • Dieckol, isolated from the edible brown algae Ecklonia cava, induces apoptosis of ovarian cancer cells and inhibits tumor xenograft growth.
Dieckol, isolated from the edible brown algae Ecklonia cava, induces apoptosis of ovarian cancer cells and inhibits tumor xenograft growth.
Journal of cancer research and clinical oncology (2014-09-14)
Ji-Hye Ahn, Yeong-In Yang, Kyung-Tae Lee, Jung-Hye Choi
ABSTRAKT
Ecklonia cava is an abundant brown alga and has been reported to possess various bioactive compounds having anti-inflammatory effect. However, the anticancer effects of dieckol, a major active compound in E. cava, are poorly understood. In the present study, we investigated the anti-tumor activity of dieckol and its molecular mechanism in ovarian cancer cells and in a xenograft mouse model . MTT assay, PI staining, and PI and Annexin double staining were performed to study cell cytotoxicity, cell cycle distribution, and apoptosis. We also investigated reactive oxygen species (ROS) production and protein expression using flow cytometry and Western blot analysis, respectively. Anti-tumor effects of dieckol were evaluated in SKOV3 tumor xenograft model. We found that the E. cava extract and its phlorotannins have cytotoxic effects on A2780 and SKOV3 ovarian cancer cells. Dieckol induced the apoptosis of SKOV3 cells and suppressed tumor growth without any significant adverse effect in the SKOV3-bearing mouse model. Dieckol triggered the activation of caspase-8, caspase-9, and caspase-3, and pretreatment with caspase inhibitors neutralized the pro-apoptotic activity of dieckol. Furthermore, treatment with dieckol caused mitochondrial dysfunction and suppressed the levels of anti-apoptotic proteins. We further demonstrated that dieckol induced an increase in intracellular ROS, and the antioxidant N-acetyl-L-cysteine (NAC) significantly reversed the caspase activation, cytochrome c release, Bcl-2 downregulation, and apoptosis that were caused by dieckol. Moreover, dieckol inhibited the activity of AKT and p38, and overexpression of AKT and p38, at least in part, reversed dieckol-induced apoptosis in SKOV3 cells. These data suggest that dieckol suppresses ovarian cancer cell growth by inducing caspase-dependent apoptosis via ROS production and the regulation of AKT and p38 signaling.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Dihydroethidium, ≥95%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Dihydroethidium, BioReagent, suitable for fluorescence, ≥95% (HPCE)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
N-Acetyl-Leu-Glu-His-Asp-7-amido-4-trifluoromethylcoumarin, ≥95% (HPLC), powder
Sigma-Aldrich
Propidium iodide solution
USP
Alkohol metylowy, United States Pharmacopeia (USP) Reference Standard