Merck
Critical role of neutral cholesteryl ester hydrolase 1 in cholesteryl ester hydrolysis in murine macrophages.
Journal of lipid research (2014-05-29)
Kent Sakai, Masaki Igarashi, Daisuke Yamamuro, Taichi Ohshiro, Shuichi Nagashima, Manabu Takahashi, Bolormaa Enkhtuvshin, Motohiro Sekiya, Hiroaki Okazaki, Jun-ichi Osuga, Shun Ishibashi
ABSTRAKT
Hydrolysis of intracellular cholesteryl ester (CE) is the rate-limiting step in the efflux of cholesterol from macrophage foam cells. In mouse peritoneal macrophages (MPMs), this process is thought to involve several enzymes: hormone-sensitive lipase (Lipe), carboxylesterase 3 (Ces3), neutral CE hydrolase 1 (Nceh1). However, there is some disagreement over the relative contributions of these enzymes. To solve this problem, we first compared the abilities of several compounds to inhibit the hydrolysis of CE in cells overexpressing Lipe, Ces3, or Nceh1. Cells overexpressing Ces3 had negligible neutral CE hydrolase activity. We next examined the effects of these inhibitors on the hydrolysis of CE and subsequent cholesterol trafficking in MPMs. CE accumulation was increased by a selective inhibitor of Nceh1, paraoxon, and two nonselective inhibitors of Nceh1, (+)-AS115 and (-)-AS115, but not by two Lipe-selective inhibitors, orlistat and 76-0079. Paraoxon inhibited cholesterol efflux to apoA-I or HDL, while 76-0079 did not. These results suggest that Nceh1 plays a dominant role over Lipe in the hydrolysis of CE and subsequent cholesterol efflux in MPMs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Oleic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Paraoxon-ethyl, PESTANAL®, analytical standard
Sigma-Aldrich
4-Nitrophenyl butyrate, ≥98%
Sigma-Aldrich
Paraoxon-ethyl, ≥90%, oil
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, technical grade, 90%
Supelco
Oleic acid, Selectophore, ≥99%
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture