Merck
A polymeric conjugate foreignizing tumor cells for targeted immunotherapy in vivo.
Journal of controlled release : official journal of the Controlled Release Society (2014-12-17)
Young-Ho Lee, Hong Yeol Yoon, Jung Min Shin, G Saravanakumar, Kyung Hee Noh, Kwon-Ho Song, Ju-Hong Jeon, Dong-Wan Kim, Kyung-Mi Lee, Kwangmeyung Kim, Ick Chan Kwon, Jae Hyung Park, Tae Woo Kim
ABSTRAKT
Antigen-specific CD8(+) cytotoxic T lymphocytes (CTLs) are key elements of immunological rejection in transplantation as well as cancer immunotherapy. Most tumors, however, are not immunologically rejected because they have self antigens, which are not recognized as the foreigner by CTLs. In this study, we hypothesized that "foreignizing" tumor cells by delivering non-self foreign antigens into the tumors would result in rejection by foreign antigen-reactive CTLs. As the model system to foreignize the tumors, we prepared a polymeric conjugate consisting of hyaluronic acid as the CD44(+) tumor-targeting ligand and ovalbumin (OVA) as a foreign antigen. When the conjugate was treated with CD44(high) TC-1 tumor cells, it was effectively taken up and allowed for displaying of antigenic OVA257-264 peptide at MHC class I on the surface of the cells. In addition, the conjugate was effectively accumulated into tumor tissue after its systemic administration to mice which are immunized with a vaccine for a vaccinia virus expressing OVA to generate OVA257-264 specific CTLs, resulting in substantial inhibition of tumor growth. Overall, these results suggest that the polymeric conjugates bearing foreign antigens may be innovative and promising cancer immunotherapeutic agents by foreignizing tumor cells, leading to immunological rejection.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium cyanoborohydride, reagent grade, 95%
Sigma-Aldrich
Sodium cyanoborohydride solution, 1.0 M in THF
Sigma-Aldrich
Sodium cyanoborohydride solution, 5.0 M in 1 M NaOH
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)