Merck
Modelling Fanconi anemia pathogenesis and therapeutics using integration-free patient-derived iPSCs.
Nature communications (2014-07-08)
Guang-Hui Liu, Keiichiro Suzuki, Mo Li, Jing Qu, Nuria Montserrat, Carolina Tarantino, Ying Gu, Fei Yi, Xiuling Xu, Weiqi Zhang, Sergio Ruiz, Nongluk Plongthongkum, Kun Zhang, Shigeo Masuda, Emmanuel Nivet, Yuji Tsunekawa, Rupa Devi Soligalla, April Goebl, Emi Aizawa, Na Young Kim, Jessica Kim, Ilir Dubova, Ying Li, Ruotong Ren, Chris Benner, Antonio Del Sol, Juan Bueren, Juan Pablo Trujillo, Jordi Surralles, Enrico Cappelli, Carlo Dufour, Concepcion Rodriguez Esteban, Juan Carlos Izpisua Belmonte
ABSTRAKT
Fanconi anaemia (FA) is a recessive disorder characterized by genomic instability, congenital abnormalities, cancer predisposition and bone marrow (BM) failure. However, the pathogenesis of FA is not fully understood partly due to the limitations of current disease models. Here, we derive integration free-induced pluripotent stem cells (iPSCs) from an FA patient without genetic complementation and report in situ gene correction in FA-iPSCs as well as the generation of isogenic FANCA-deficient human embryonic stem cell (ESC) lines. FA cellular phenotypes are recapitulated in iPSCs/ESCs and their adult stem/progenitor cell derivatives. By using isogenic pathogenic mutation-free controls as well as cellular and genomic tools, our model serves to facilitate the discovery of novel disease features. We validate our model as a drug-screening platform by identifying several compounds that improve hematopoietic differentiation of FA-iPSCs. These compounds are also able to rescue the hematopoietic phenotype of FA patient BM cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Ganciclovir, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Ganciclovir, ≥99% (HPLC), powder
Sigma-Aldrich
1,3-Butadiene diepoxide, 97%
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Proline, European Pharmacopoeia (EP) Reference Standard
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Fluorescein, European Pharmacopoeia (EP) Reference Standard
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
Supelco
Resveratrol, analytical standard
Supelco
Resveratrol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Sigma-Aldrich
Anti-β-Tubulin III antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Supelco
Sodium butyrate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Sodium butyrate, 98%
Sigma-Aldrich
DL-Proline, ReagentPlus®, 99%
Sigma-Aldrich
Danazol, ≥98%
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Fluorescein, free acid
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder