Merck
Dual receptor-specific peptides modified liposomes as VEGF siRNA vector for tumor-targeting therapy.
Current gene therapy (2014-07-22)
Zhenzhen Yang, Bai Xiang, Dawen Dong, Zhanzhang Wang, Jingquan Li, Xianrong Qi
ABSTRAKT
Tumor angiogenesis involves multiple signaling pathways that provide potential therapeutic targets to inhibit tumor growth and metastasis. Regarding the significant role of vascular endothelial growth factor (VEGF) in angiogenesis and tumor progression, VEGF sequence-specific small interfering RNA (siRNA) for anti-angiogenic tumor therapy are under development. In the present study, dual-modified liposomes (At-Lp) was designed by attaching two receptorspecific peptides, Angiopep and tLyP-1, which specifically targeting low-density lipoprotein receptor (LRP) for brain tumor targeting and neuropilin-1 receptor (NRP-1) for tumor penetration, respectively. Gene transfection and silencing, and antitumor effect of the At-Lp loaded with VEGF siRNA were evaluated in vitro and in orthotopic xenograft models of U87 MG tumor. The At-Lp significantly enhanced cellular uptake (2-fold) and down-regulated expression of VEGF in U87 MG glioblastoma cells compared with non-modified and single-modified liposomes. The internalization of the At-Lp into tumor cells was taken via the enhanced permeability and retention effect and receptor-mediated endocytosis, followed by an effective endosomal escape of loaded siRNA into the cytoplasm. The At-Lp showed great superiority in inhibition of tumor growth, anti-angiogenesis, expression of VEGF and apoptosis effect after in vivo application against nude mice bearing U87 MG glioblastoma without activation of system-associated toxicity and the innate immune response. These results demonstrated that the combination of two receptor-specific peptides-mediated liposomes presented a promising platform for effective targeting delivery of siRNA for cancer anti-angiogenic therapy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Sulforhodamine B, Dye content 75 %
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
MISSION® esiRNA, targeting human VEGFA
Sigma-Aldrich
Mouse IL-6 R ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Rat IL-6 ELISA Kit, for serum, plasma and cell culture supernatants
Sigma-Aldrich
Rat Interleukin-6 ELISA Kit, for for cell lysate and tissue lysate
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Vegfa
Sigma-Aldrich
Sulforhodamine B sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sulforhodamine B sodium salt, Technical grade