Merck
Chitosan gel formulations containing egg yolk oil and epidermal growth factor for dermal burn treatment.
Die Pharmazie (2015-05-23)
E Yenilmez, E Başaran, R Arslan, M S Berkman, U M Güven, C Bayçu, Y Yazan
ABSTRAKT
In the present study chitosan based gel formulations containing Egg Yolk Oil (EYO) and Epidermal Growth Factor (EGF) were formulated successfully aiming at enhanced topical treatment of dermal burns the combination of traditional approaches with modern drug delivery systems. Physicochemical properties of the formulations were analyzed and efficacy of the formulations prepared were evaluated versus a commercial product; Silverdin (1% silver sulfadiazine) in vivo on Wistar rats. Burns were generated on the back of the rats and at predetermined time intervals tissue samples were collected and evaluated histologically. The analyses showed that chitosan based gel formulations containing Egg Yolk Oil (E1) and chitosan based gel formulations containing EYO and EGF (M1) formulations seem to be better alternatives for Silverdin with a significant difference (p < 0.05) considering healing ranks of tissue samples.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Xylazine hydrochloride, ≥99.0% (HPLC)
Xylazine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
USP
Kwas octowy lodowaty, United States Pharmacopeia (USP) Reference Standard