Merck
  • Immediate enhancement of first-phase insulin secretion and unchanged glucose effectiveness in patients with type 2 diabetes after Roux-en-Y gastric bypass.
Immediate enhancement of first-phase insulin secretion and unchanged glucose effectiveness in patients with type 2 diabetes after Roux-en-Y gastric bypass.
American journal of physiology. Endocrinology and metabolism (2015-01-30)
Christoffer Martinussen, Kirstine N Bojsen-Møller, Carsten Dirksen, Siv H Jacobsen, Nils B Jørgensen, Viggo B Kristiansen, Jens J Holst, Sten Madsbad
ABSTRAKT
Roux-en-Y gastric bypass surgery (RYGB) in patients with type 2 diabetes often leads to early disease remission, and it is unknown to what extent this involves improved pancreatic β-cell function per se and/or enhanced insulin- and non-insulin-mediated glucose disposal (glucose effectiveness). We studied 30 obese patients, including 10 with type 2 diabetes, 8 with impaired glucose tolerance, and 12 with normal glucose tolerance before, 1 wk, and 3 mo after RYGB, using an intravenous glucose tolerance test (IVGTT) to estimate first-phase insulin response, insulin sensitivity (Si), and glucose effectiveness with Bergman's minimal model. In the fasting state, insulin sensitivity was estimated by HOMA-S and β-cell function by HOMA-β. Moreover, mixed-meal tests and oral GTTs were performed. In patients with type 2 diabetes, glucose levels normalized after RYGB, first-phase insulin secretion in response to iv glucose increased twofold, and HOMA-β already improved 1 wk postoperatively, with further enhancements at 3 mo. Insulin sensitivity increased in the liver (HOMA-S) at 1 wk and at 3 mo in peripheral tissues (Si), whereas glucose effectiveness did not improve significantly. During oral testing, GLP-1 responses and insulin secretion increased regardless of glucose tolerance. Therefore, in addition to increased insulin sensitivity and exaggerated postprandial GLP-1 levels, diabetes remission after RYGB involves early improvement of pancreatic β-cell function per se, reflected in enhanced first-phase insulin secretion to iv glucose and increased HOMA-β. A major role for improved glucose effectiveness after RYGB was not supported by this study.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder