Merck
A new role for monocytes in modulating myometrial inflammation during human labor.
Biology of reproduction (2014-05-16)
Khetsopon Srikhajon, Oksana Shynlova, Anyarin Preechapornprasert, Boonsri Chanrachakul, Stephen Lye
ABSTRAKT
Here we fully characterize the cytokine profile of laboring human myometrium using Luminex analysis of 48 cytokine proteins, and stereologically quantified infiltration of monocytes and neutrophils into the myometrium. We hypothesized that monocytes can regulate their accumulation in the myometrium by disruption of proinflammatory cytokines to prevent an uncontrolled inflammatory response after labor onset. We isolated primary human myometrial cells (HMCs) from term, nonlaboring myometrial biopsies. Confluent HMCs were cocultured directly with human monocytic (THP-1) or lymphocytic (U937) cells, and with the same cells spatially separated by a membrane insert. After 72 h, HMCs and THP-1 were harvested separately, and RNA was extracted and analyzed by quantitative PCR. Coculture supernatants were collected and analyzed by Luminex assay and ELISA. We found that the laboring human myometrium produces significantly higher amounts of interleukin (IL) 6, IL9, IL18, IL1RA, CCL2, CCL7, CXCL8, CSF3, and tumor necrosis factor alpha, which coincides with the influx of immune cells. The direct contact or presence of THP-1 monocytes (but not U937 cells) significantly decreased CCL2 protein levels and increased IL1RA protein levels secreted by HMCs. This time-dependent decrease of CCL2 was greater with increasing number of monocytes being in direct contact with HMCs. We suggest a novel mechanism by which monocytes are first recruited to the myometrium by multiple cytokines and contribute to the physiologic inflammation of labor. After completing transmigration, activated monocytes disrupt locally established CCL2 gradients (possible by CCR2-mediated consumption) to limit their accumulation in the uterus. This mechanism may serve as a negative feedback loop to control the local inflammation and promote a return to homeostasis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
USP
Phenol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phenol, natural, 97%, FG
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology