Merck
Carbonic anhydrase IX is associated with early pulmonary spreading of primary colorectal carcinoma and tobacco smoking.
European journal of cardio-thoracic surgery : official journal of the European Association for Cardio-thoracic Surgery (2013-12-11)
Thomas Schweiger, Dagmar Kollmann, Christoph Nikolowsky, Denise Traxler, Emmanuella Guenova, György Lang, Peter Birner, Walter Klepetko, Hendrik Jan Ankersmit, Konrad Hoetzenecker
ABSTRAKT
Pulmonary metastasectomy is performed routinely in selected patients with metastatic spreading to the lungs. According to current guidelines, the tumour biology should be taken into account when selecting patients for a resection. Carbonic anhydrase IX (CA9) expression has been shown to be a common feature in primary tumour growth and metastasis and it negatively affects the clinical outcome in various malignancies. Data on CA9 in pulmonary metastases are lacking. Forty-four patients with primary colorectal cancer (CRC) who underwent curative pulmonary metastasectomy were included in this study. We determined the expression of CA9 in pulmonary metastases and corresponding primaries by immunohistochemistry. The expression level was correlated with clinical parameters and patients' smoking habits. Furthermore, the impact of nicotine treatment on phosphorylation of STAT3, HIF-1α and CA9 expression was assessed in HT29 cells. High expression of CA9 in resected pulmonary metastases and corresponding primary tumours correlated with early spreading to the lung (both P < 0.001). Moreover, CA9 expression was affected by the smoking habits of the patients. Treating HT29 cells with nicotine resulted in an induction of CA9 in vitro. This induction was associated with STAT3 phosphorylation and was independent of HIF-1α. This study provides first evidence of CA9 expression in pulmonary metastases of CRC and suggests a role of CA9 as a prognostic marker. Moreover, our in vitro and in vivo data indicate an association between tobacco smoking and CA9 expression. Immunohistochemical assessment of CA9 expression might serve as an additional tool in decision-making for selecting patients for pulmonary metastasectomy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
(−)-Nicotine, PESTANAL®, analytical standard
Sigma-Aldrich
(−)-Nicotine, ≥99% (GC), liquid
Sigma-Aldrich
(±)-Nicotine, ≥99% (TLC), liquid
Sigma-Aldrich
Human Carbonic Anhydrase IX ELISA Kit, for serum, plasma, cell culture supernatant and urine