Merck
Development and validation of a rapid HPLC method for the quantification of GSE4 peptide in biodegradable PEI-PLGA nanoparticles.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-12-03)
Susana P Egusquiaguirre, Cristina Manguán-García, Rosario Perona, José Luís Pedraz, Rosa Maria Hernández, Manuela Igartua
ABSTRAKT
In this work a high performance liquid chromatographic (HPLC) method has been developed and validated for the content determination of GSE4 peptide in PEI-PLGA nanoparticles. Chromatographic separation was performed on a C18 column, and a gradient elution with a mobile phase composed of methanol and 0.1% aqueous trifluoroacetic acid (TFA) solution, at a flow rate of 1ml/min, was used. GSE4 peptide identification was made by fluorescence detection at 290nm. The elution of methanol:TFA was initially maintained at (20:80, v/v) for one min and the gradient changed to (80:20, v/v) in 6min. This ratio was then followed by isocratic elution at (80:20, v/v) during another min and for further 3min it was linearly modified to (20:80, v/v). The developed method was validated according to the ICH guidelines, being specific, linear in the range 10-100μg/ml (R(2)=0.9996), precise, exhibiting good inter-day and intra-day precision reflected by the relative standard deviation values (less than 3.88%), accurate, with a recovery rate of 100.18±0.95%, and stable for 48h at 5°C or at RT when encapsulated in nanoparticles. The method was simple, fast, and successfully used to determine the peptide content in GSE4-loaded PEI-PLGA nanoparticles.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
USP
Alkohol metylowy, United States Pharmacopeia (USP) Reference Standard