Merck
Registered report: Oncometabolite 2-hydroxyglutarate is a competitive inhibitor of α-ketoglutarate-dependent dioxygenases.
eLife (2015-08-01)
Brad Evans, Erin Griner
ABSTRAKT
The Reproducibility Project: Cancer Biology seeks to address growing concerns about reproducibility in scientific research by conducting replications of selected experiments from a number of high-profile papers in the field of cancer biology. The papers, which were published between 2010 and 2012, were selected on the basis of citations and Altmetric scores (Errington et al., 2014). This Registered report describes the proposed replication plan of key experiments from 'Oncometabolite 2-hydroxyglutarate is a competitive inhibitor of α-ketoglutarate-dependent dioxygenases' by Xu and colleagues, published in Cancer Cell in 2011 (Xu et al., 2011). The key experiments being replicated include Supplemental Figure 3I, which demonstrates that transfection with mutant forms of IDH1 increases levels of 2-hydroxyglutarate (2-HG), Figures 3A and 8A, which demonstrate changes in histone methylation after treatment with 2-HG, and Figures 3D and 7B, which show that mutant IDH1 can effect the same changes as treatment with excess 2-HG. The Reproducibility Project: Cancer Biology is a collaboration between the Center for Open Science and Science Exchange, and the results of the replications will be published by eLife.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Tris Buffered Saline, 10 ×, solution
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
D-α-Hydroxyglutaric acid disodium salt, ≥95% (GC)
Informacje o cenach i dostępności nie są obecnie dostępne.
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
L-α-Hydroxyglutaric acid disodium salt, ≥98.0% (GC)
Informacje o cenach i dostępności nie są obecnie dostępne.