Merck
Antiplatelet and antithrombotic activity of a fibrin(ogen)olytic protease from Bacillus cereus strain FF01.
International journal of biological macromolecules (2015-05-13)
Sourav Majumdar, Sumita Dutta, Tanusree Das, Pronobesh Chattopadhyay, Ashis K Mukherjee
ABSTRAKT
Fibrin(ogen)olytic enzymes offer great promise for the treatment of thrombosis associated disorders. The present study describes the characterization of an extracellular fibrin(ogen)olytic serine protease (named Bacethrombase) purified from the Bacillus cereus strain FF01. The molecular mass of the Bacethrombase was determined by sodium dodecyl sulfate polyacrylamide gel electrophoresis and matrix assisted laser desorption/ionization-time-of-flight-mass spectroscopy analyses at 39.5 kDa and 38,450.51 Da, respectively. The peptide mass fingerprinting and analyses of the composition of the amino acids revealed the similarity of the Bacethrombase to the bacterial serine proteases. The secondary structure of the Bacethrombase was composed of 14% helix, 6.6% beta-sheet, and 79.4% random coil. Bacethrombase was found to contain 48% sialic acid and it preferentially degraded the Aα-chain of fibrinogen, as well as fibrin. The anticoagulant potency of the Bacethrombase was comparable with that of warfarin and heparin, and was corroborated by its fibrinogenolytic activity rather than the inhibition of thrombin, prothrombin or FXa. Bacethrombase demonstrated antiplatelet activity, and dose-dependently inhibited the ADP-induced platelet aggregation. Bacethrombase (10 mg/kg) did not show toxicity after i.v. administration in Wistar rats; however, it revealed an in vivo anticoagulant effect and significantly inhibited the carrageenan-induced in vivo thrombus formation in rats.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ammonium-14N2,sulfate-16O4, 99.99 atom % 16O, 99.99 atom % 14N
Sigma-Aldrich
3-Mercaptopropionic acid, ≥99%
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Sinapic acid, ≥98%, powder
Sigma-Aldrich
3-Mercaptopropionic acid, ≥99.0% (HPLC)
Sigma-Aldrich
Ammonium-14N2 sulfate, 99.99 atom % 14N
Sigma-Aldrich
Fmoc chloride, 97%
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
D-Val-Leu-Lys 4-nitroanilide dihydrochloride, plasmin substrate
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, Molecular Biology, ≥99.0%
Sigma-Aldrich
Fmoc chloride, BioReagent, ≥99.0% (HPLC)
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis