Merck
Endocrine-disrupting effect of the ultraviolet filter benzophenone-3 in zebrafish, Danio rerio.
Environmental toxicology and chemistry (2015-06-30)
Karin L Kinnberg, Gitte I Petersen, Mette Albrektsen, Mita Minghlani, Suad Mohamud Awad, Bente F Holbech, John W Green, Poul Bjerregaard, Henrik Holbech
ABSTRAKT
The chemical ultraviolet (UV) filter benzophenone-3 (BP-3) is suspected to be an endocrine disruptor based on results from in vitro and in vivo testing. However, studies including endpoints of endocrine adversity are lacking. The present study investigated the potential endocrine-disrupting effects of BP-3 in zebrafish (Danio rerio) in the Fish Sexual Development Test (Organisation for Economic Co-operation and Development TG 234) and a 12-d adult male zebrafish study. In TG 234, exposure from 0 d to 60 d posthatch caused a monotone dose-dependent skewing of the phenotypic sex ratio toward fewer males and more female zebrafish (no observed effect concentration [NOEC]: 191 μg/L, lowest observed effect concentration [LOEC]: 388 μg/L). Besides, gonad maturation was affected in both female fish (NOEC 191 μg/L, LOEC 388 μg/L) and male fish (NOEC 388 μg/L, LOEC 470 μg/L). Exposure to BP-3 did not affect the vitellogenin concentration in TG 234. After 12 d exposure of adult male zebrafish, a slight yet significant increase in the vitellogenin concentration was observed at 268 μg/L but not at 63 μg/L and 437 μg/L BP-3. Skewing of the sex ratio is a marker of an endocrine-mediated mechanism as well as a marker of adversity, and therefore the conclusion of the present study is that BP-3 is an endocrine-disrupting chemical in accordance with the World Health Organization's definition.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Supelco
17β-Estradiol solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Hydroxy-4-methoxybenzophenone, 98%
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Isopropanol, 70% in H2O