Merck
  • In vitro activity of colistin in antimicrobial combination against carbapenem-resistant Acinetobacter baumannii isolated from patients with ventilator-associated pneumonia in Vietnam.
In vitro activity of colistin in antimicrobial combination against carbapenem-resistant Acinetobacter baumannii isolated from patients with ventilator-associated pneumonia in Vietnam.
Journal of medical microbiology (2015-08-25)
Vien Le Minh, Nguyen Thi Khanh Nhu, Voong Vinh Phat, Corinne Thompson, Nguyen Phu Huong Lan, Tran Vu Thieu Nga, Pham Thi Thanh Tam, Ha Thanh Tuyen, Tran Do Hoang Nhu, Nguyen Van Hao, Huynh Thi Loan, Lam Minh Yen, Christopher M Parry, Ho Dang Trung Nghia, James I Campbell, Tran Tinh Hien, Louise Thwaites, Guy Thwaites, Nguyen Van Vinh Chau, Stephen Baker
ABSTRAKT
Acinetobacter baumannii has become one of the major infection threats in intensive care units (ICUs) globally. Since 2008, A. baumannii has been the leading cause of ventilator-associated pneumonia (VAP) in our ICU at an infectious disease hospital in southern Vietnam. The emergence of this pathogen in our setting is consistent with the persistence of a specific clone exhibiting resistance to carbapenems. Antimicrobial combinations may be a strategy to treat infections caused by these carbapenem-resistant A. baumannii. Therefore, we assessed potential antimicrobial combinations against local carbapenem-resistant A. baumannii by measuring in vitro interactions of colistin with four antimicrobials that are locally certified for treating VAP. We first performed antimicrobial susceptibility testing and multilocus variable number tandem repeat analysis (MLVA) genotyping on 74 A. baumannii isolated from quantitative tracheal aspirates from patients with VAP over an 18-month period. These 74 isolates could be subdivided into 21 main clusters by MLVA and >80 % were resistant to carbapenems. We selected 56 representative isolates for in vitro combination synergy testing. Synergy was observed in four (7 %), seven (13 %), 20 (36 %) and 38 (68 %) isolates with combinations of colistin with ceftazidime, ceftriaxone, imipenem and meropenem, respectively. Notably, more carbapenem-resistant A. baumannii isolates (36/43; 84 %) exhibited synergistic activity with a combination of colistin and meropenem than carbapenem-susceptible A. baumannii isolates (2/13; 15 %) (P = 0.023; Fisher's exact test). Our findings suggest that combinations of colistin and meropenem should be considered when treating carbapenem-resistant A. baumannii infections in Vietnam, and we advocate clinical trials investigating combination therapy for VAP.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Formamide, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Formamide, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Formamide, spectrophotometric grade, ≥99%
Sigma-Aldrich
Formamide, ≥99.5% (GC), BioReagent, Molecular Biology
Supelco
Formamide solution, suitable for NMR (reference standard), 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Formamide solution, suitable for NMR (reference standard), 90% in DMSO-d6 (99.9 atom % D), NMR tube size 10 mm × 8 in.
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis