Merck
Antibacterial carboxymethyl cellulose/Ag nanocomposite hydrogels cross-linked with layered double hydroxides.
International journal of biological macromolecules (2015-05-13)
Mehdi Yadollahi, Hassan Namazi, Mohammad Aghazadeh
ABSTRAKT
This paper deals with the preparation of antibacterial nanocomposite hydrogels through the combination of carboxy methyl cellulose (CMC), layered double hydroxides (LDH), and silver nanoparticles (AgNPs). CMC-LDH hydrogels were prepared by intercalating CMC into different LDHs. Then, Ag/CMC-LDH nanocomposite hydrogels were prepared through in situ formation of AgNPs within the CMC-LDHs. XRD analysis confirmed the intercalating CMC into the LDH sheets and formation of intercalated structures, as well as formation of AgNPs within the CMC-LDHs. SEM and TEM micrographs indicated well distribution of AgNPs within the Ag/CMC-LDHs. The prepared hydrogels showed a pH sensitive swelling behavior. The Ag/CMC-LDH nanocomposite hydrogels have rather higher swelling in different aqueous solutions in comparison with CMC-LDHs. The antibacterial activity of CMC-LDHs increased considerably after formation of AgNPs and was stable for more than one month.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Sigma-Aldrich
Silver nitrate, ≥99.999% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O
Sigma-Aldrich
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh