Merck
  • Electrochemical immunosensor for sensitive determination of the anorexigen peptide YY at grafted reduced graphene oxide electrode platforms.
Electrochemical immunosensor for sensitive determination of the anorexigen peptide YY at grafted reduced graphene oxide electrode platforms.
The Analyst (2015-08-11)
S Guerrero, G Martínez-García, V Serafín, L Agüí, P Yáñez-Sedeño, J M Pingarrón
ABSTRAKT
The first electrochemical immunosensor for the determination of peptide YY is reported in this paper. A novel electrochemical platform, prepared by the electrochemical grafting of the diazonium salt of 4-aminobenzoic acid onto a reduced graphene oxide-modified glassy carbon electrode, was used, on which the covalent immobilization of specific anti-PYY antibodies was accomplished. The HOOC-Phe-rGO/GCEs were characterized using cyclic voltammetry and electrochemical impedance spectroscopy. The different variables affecting the preparation of the modified electrodes and the performance of the immunosensor were optimized. Under the optimized conditions, a calibration plot for PYY showing a linear range extending between 10(-4) and 10(2) ng mL(-1) was found. This range is adequate for the determination of this protein in real samples, since the expected concentration in human serum is around 100 pg mL(-1). The limit of detection was 0.01 pg mL(-1) of PYY. The immunosensor exhibited good reproducibility of the PYY measurements, excellent storage stability and selectivity, as well as a shorter assay time than those of ELISA kits. The usefulness of the immunosensor for the analysis of real samples was demonstrated by analyzing human serum samples spiked with PYY at three concentration levels.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Ethanolamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
1-Naphthyl phosphate, 99%
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
(+)-Abscisic acid, ≥98% (HPLC)
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
4-Aminobenzoic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
4-Aminobenzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
4-Aminobenzoic acid, purified by sublimation, ≥99%
Sigma-Aldrich
Potassium chloride, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Potassium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, Molecular Biology, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Ethanolamine, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethanolamine, ≥98%
Sigma-Aldrich
Ethanolamine, ≥99%
Sigma-Aldrich
Ethanolamine, liquid, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Sodium phosphate dibasic, Molecular Biology, ≥98.5% (titration)