Merck
  • Evidence of Sulfur Mustard Exposure in Human Plasma by LC-ESI-MS-MS Detection of the Albumin-Derived Alkylated HETE-CP Dipeptide and Chromatographic Investigation of Its Cis/Trans Isomerism.
Evidence of Sulfur Mustard Exposure in Human Plasma by LC-ESI-MS-MS Detection of the Albumin-Derived Alkylated HETE-CP Dipeptide and Chromatographic Investigation of Its Cis/Trans Isomerism.
Journal of analytical toxicology (2015-02-26)
Felix Gandor, Michael Gawlik, Horst Thiermann, Harald John
ABSTRAKT
Sulfur mustard (SM) is a chemical warfare agent that causes painful blisters and chemically modifies endogenous biomacromolecules by alkylation to hydroxyethylthioethyl (HETE) adducts representing valuable long-term markers for post-exposure analysis. The albumin adduct formed in human plasma in vitro (HETE bound to the side chain of cysteine 34) was isolated and cleaved by current lots of pronase primarily generating the internal modified dipeptide (HETE-cysteine-proline, HETE-CP) instead of the formerly reported HETE-CPF tripeptide. The analyte was detected by liquid chromatography-electrospray ionization tandem-mass spectrometry (LC-ESI-MS-MS). In principle, HETE-CP undergoes a dynamic on-column equilibrium of cis-trans isomerism thus requiring separation at 50°C to obtain one narrow peak. Accordingly, we developed both a novel longer lasting but more sensitive microbore (1 mm i.d., flow 30 µL/min, cycle time 60 min, LOD 50 nM) and a faster, less sensitive narrowbore (2.1 mm i.d., 200 µL/min, cycle time 16 min, LOD 100 nM, both on Atlantis T3 material at 50°C) LC-ESI-MS-MS method suitable for verification analysis. The corresponding tri- and tetrapeptide, Q(HETE)-CPF were monitored simultaneously. HETE-CP peak areas were directly proportional to SM concentrations added to plasma in vitro (0.05-100 µM). Albumin adducts formed by deuterated SM (d8-SM) served as internal standard.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis