Merck
The acetylcholinesterase (AChE) inhibition analysis of medaka (Oryzias latipes) in the exposure of three insecticides.
Pakistan journal of pharmaceutical sciences (2015-03-23)
Jianping Zhu, Cheng Huan, Guiyun Si, Haitang Yang, Li Yin, Qing Ren, Baixiang Ren, Rongshu Fu, Mingsheng Miao, Zongming Ren
ABSTRAKT
The continuous effects on Acetylcholinesterase (AChE) activity of medaka (Oryzias latipes) caused by dichlorvos, methomyl and deltamethrin in vivo were investigated, and the trends of AChE activity inhibition due to the influence of these insecticides were discussed. The LC50-24h of dichlorvos, methomyl and deltamethrin on medaka were 2.3 mg/L, 0.2 mg/L, and 2.9×10(-3) mg/L respectively. The result suggested that at the beginning of the exposure, the AChE activity might increase, and the AChE activity in dead individuals was obviously lower than the live individuals. Though the de novo synthesis of AChE in medaka might help the AChE activity recover, the trends during the exposure in different treatments were downward, and it showed both exposure time and concentration dependent. Meanwhile, higher temperature might cause the AChE inhibition earlier due to the higher metabolic rate. Therefore, as a specific biomarker for organophosphate, carbamate pesticides and pyrethroids, the degree of the AChE inhibition with in vivo conditions is a good tool in continuous monitoring of insecticides, which may induce the nerve conduction disorders.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
2-Nitrobenzoic acid, 95%, Contains 3- and 4-isomers
Sigma-Aldrich
Acetylthiocholine iodide, ≥99.0% (AT)
Sigma-Aldrich
Acetylthiocholine iodide, ≥98% (TLC), powder or crystals
Sigma-Aldrich
Triton X-100, laboratory grade