Merck
Towards high potential magnetic biocatalysts for on-demand elimination of pharmaceuticals.
Bioresource technology (2015-10-18)
Vaidyanathan Vinoth Kumar, Hubert Cabana
ABSTRAKT
The present study investigated the applicability of a laccase based bioprocess for the treatment of a mixture containing 13 selected pharmaceuticals. To do so, laccase was immobilized as cross-linked enzyme aggregates (MAC-CLEAs) on amine functionalized magnetic nanoparticles using chitosan/1-ethyl-3-(3-dimethylaminopropyl)-carbodiimide hydrochloride (EDAC) as the cross-linking system. The activity recovery of laccase reached 61.4% under the optimal conditions of MAC-CLEAs formation. The latter exhibited enhanced storage stability over one year at 4°C and showed better temperature resistance compared to its soluble counterpart. The biocatalysts were properly recycled and the catalytic activity recovery was good even after a hundred and fifty batch reactions. Complete removal of pharmaceuticals like acetaminophen, diclofenac, mefenamic acid, atenolol and epoxy carbamazepine and partial removal of fenofibrate, diazepam, trimethoprim, and ketoprofen by laccase was achieved within 12h of incubation, whereas efficient removal of indometacin required the presence of mediator.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Carbamazepine, powder
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder
Sigma-Aldrich
Fenofibrate, ≥99%, powder
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)
Sigma-Aldrich
Naproxen, meets USP testing specifications
Supelco
Diazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Ketoprofen, ≥98% (TLC)
Sigma-Aldrich
Ketoprofen, meets USP testing specifications
Sigma-Aldrich
(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, 98%
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, BioXtra
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, commercial grade, powder
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, ≥99.0% (AT)
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, crystalline