Merck
Nature-inspired DNA nanosensor for real-time in situ detection of mRNA in living cells.
ACS nano (2015-04-24)
Chor Yong Tay, Liang Yuan, David Tai Leong
ABSTRAKT
Rapid and precise in situ detection of gene expressions within a single cell is highly informative and offers valuable insights into its state. Detecting mRNA within single cells in real time and nondestructively remains an important challenge. Using DNA nanotechnology and inspired by nature's many examples of "protective-yet-accessible" exoskeletons, we designed our mRNA nanosensor, nano-snail-inspired nucleic acid locator (nano-SNEL), to illustrate these elements. The design of the nano-SNEL is composed of a sensory molecular beacon module to detect mRNA and a DNA nanoshell component, mimicking the functional anatomy of a snail. Accurate and sensitive visualization of mRNA is achieved by the exceptional protection conferred by the nanoshell to the sensory component from nucleases-mediated degradation by approximately 9-25-fold compared to its unprotected counterpart. Our nano-SNEL design strategy improved cell internalization is a demonstration of accurate, dynamic spatiotemporal resolved detection of RNA transcripts in living cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
Phenylacetic acid, 99%