Merck
RIP1 modulates death receptor mediated apoptosis and autophagy in macrophages.
Molecular oncology (2015-01-15)
Zhenyu Yao, Peng Zhang, Hui Guo, Juan Shi, Shilian Liu, Yanxin Liu, Dexian Zheng
ABSTRAKT
Macrophages are responsible for defending against diverse pathogens and play a crucial role in the innate immune system. Macrophage's lifespan is determined by homeostatic balance between survival and apoptosis. Here we report that tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) triggers both apoptosis and autophagy in human U937 cells. Inhibition of autophagy facilitates TRAIL-induced apoptosis, suggesting that autophagy of macrophages protects against TRAIL-induced apoptosis. TRAIL treatment influences the expression of death receptors, indicating that TRAIL-induced apoptosis and autophagy are mediated by death receptors. RIP1 ubiquitination and expression regulate apoptosis and autophagy. Furthermore, expression and bioactivity of the p43/41-caspase-8 variant are critical to TRAIL-induced autophagy and apoptosis. Knockdown of RIP1 suppresses autophagy in macrophage. These data demonstrate that RIP1 is essential for the regulation of death receptor mediated autophagy and apoptosis. The results in this study contribute to understanding the regulation of autophagy and apoptosis in macrophages, and shed lights on death receptor-targeted therapy for cancer, inflammation and autoimmune diseases.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP), meets EP testing specifications
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)