Merck
  • Toxigenic potentiality of Aspergillus flavus and Aspergillus parasiticus strains isolated from black pepper assessed by an LC-MS/MS based multi-mycotoxin method.
Toxigenic potentiality of Aspergillus flavus and Aspergillus parasiticus strains isolated from black pepper assessed by an LC-MS/MS based multi-mycotoxin method.
Food microbiology (2015-09-05)
Pratheeba Yogendrarajah, Frank Devlieghere, Emmanuel Njumbe Ediage, Liesbeth Jacxsens, Bruno De Meulenaer, Sarah De Saeger
ABSTRAKT
A liquid chromatography triple quadrupole tandem mass spectrometry method was developed and validated to determine mycotoxins, produced by fungal isolates grown on malt extract agar (MEA). All twenty metabolites produced by different fungal species were extracted using acetonitrile/1% formic acid. The developed method was applied to assess the toxigenic potentiality of Aspergillus flavus (n = 11) and Aspergillus parasiticus (n = 6) strains isolated from black peppers (Piper nigrum L.) following their growth at 22, 30 and 37 °C. Highest mean radial colony growth rates were observed at 30 °C for A. flavus (5.21 ± 0.68 mm/day) and A. parasiticus (4.97 ± 0.33 mm/day). All of the A. flavus isolates produced aflatoxin B1 and O-methyl sterigmatocystin (OMST) while 91% produced aflatoxin B2 (AFB2) and 82% of them produced sterigmatocystin (STERIG) at 30 °C. Except one, all the A. parasiticus isolates produced all the four aflatoxins, STERIG and OMST at 30 °C. Remarkably high AFB1 was produced by some A. flavus isolates at 22 °C (max 16-40 mg/kg). Production of mycotoxins followed a different trend than that of growth rate of both species. Notable correlations were found between different secondary metabolites of both species; R(2) 0.87 between AFB1 and AFB2 production. Occurrence of OMST could be used as a predictor for AFB1 production.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethanolamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Aflatoxin B1 from Aspergillus flavus, from Aspergillus flavus
Sigma-Aldrich
Ethanolamine, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethanolamine, ≥98%
Sigma-Aldrich
Ethanolamine, ≥99%
Sigma-Aldrich
Ethanolamine, liquid, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Chloramphenicol, meets USP testing specifications
Sigma-Aldrich
Chloramphenicol, BioReagent, suitable for plant cell culture
Sigma-Aldrich
T-2 Toxin, from Fusarium sp., powder, ≥98% (HPLC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)