Merck
MAPKs' status at early stages of renal carcinogenesis and tumors induced by ferric nitrilotriacetate.
Molecular and cellular biochemistry (2015-03-01)
Francisco A Aguilar-Alonso, José D Solano, Chabetty Y Vargas-Olvera, Ignacio Pacheco-Bernal, Telma O Pariente-Pérez, María Elena Ibarra-Rubio
ABSTRAKT
Renal cell carcinoma (RCC) is asymptomatic at early stages, and thus, initial diagnosis frequently occurs at advanced or even metastatic stages, leading to a high rate of mortality. Ferric nitrilotriacetate (FeNTA)-induced RCC model is a useful tool to analyze molecular events at different stages of the carcinogenesis process in vivo. MAPKs' alterations seem to play an important role in the development and maintenance of human RCC tumors. Based on the above, p38α/β/γ, JNK1/2, and ERK1/2 statuses were studied at early stages of FeNTA-induced renal carcinogenesis (1 and 2 months of carcinogen treatment) as well as in tumor tissue. MAPKs showed distinct response along carcinogenesis process, either as total proteins and/or as their phosphorylated forms. While the increase in total and phospho-p38α/β levels became lower as carcinogenesis progressed, p38γ overexpression grew. Instead, total JNK2 diminished, but JNK1 was elevated at all studied times, and p-JNK1 levels increased at early stages, but not in tumors. In contrast, p-JNK2 rose at 2 months of treatment and in tumor tissue. Increased levels of p-ERK1/2 were observed at all stages analyzed. Very interestingly, at 1 and 2 months of FeNTA treatment, no alterations in MAPKs were found in liver or lung, where no primary tumors are induced with the scheme of FeNTA administration followed here. In conclusion, MAPKs' behavior evolved differentially as renal carcinogenesis advanced, even among isoforms of the same family, but it did not change in other tissues. All this strongly suggests a role of these kinases in FeNTA-induced RCC tumor development and maintenance.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder