Merck
Combined targeting of high-mobility group box-1 and interleukin-8 to control micrometastasis potential in gastric cancer.
International journal of cancer (2015-03-31)
Hye Won Chung, Sunphil Jang, Hoguen Kim, Jong-Baeck Lim
ABSTRAKT
Micrometastasis is the major cause of treatment failure in gastric cancer (GC). Because epithelial-to-mesenchymal transition (EMT) is considered to develop prior to macroscopic metastasis, EMT-promoting factors may affect micrometastasis. This study aimed to evaluate the role of extracellular high-mobility group box-1 (HMGB1) in EMT and the treatment effect of combined targeting of HMGB1 and interleukin-8 (IL-8) at early-stage GC progression through interrupting EMT promotion. Extracellular HMGB1 was induced by human recombinant HMGB1 and pCMV-SPORT6-HMGB1 plasmid transfection. EMT activation was evaluated by immunoblotting, immunofluorescence and immunohistochemistry. Increased migration/invasion activities were evaluated by in vitro transwell migration/invasion assay using all histological types of human GC cell lines (N87, MKN28 SNU-1 and KATOIII), N87-xenograft BALB/c nude mice and human paired serum-tissue GC samples. HMGB1-induced soluble factors were measured by chemiluminescent immunoassay. Inhibition effects of tumor growth and EMT activation by combined targeting of HMGB1 and IL-8 were evaluated in N87-xenograft nude mice. Serum HMGB1 increases along the GC carcinogenesis and reaches maximum before macroscopic metastasis. Overexpressed extracellular HMGB1 promoted EMT activation and increased cell motility/invasiveness through ligation to receptor for advanced glycation end products. HMGB1-induced IL-8 overexpression contributed the HMGB1-induced EMT in GC in vitro and in vivo. Blocking HMGB1 caused significant reduction of tumor growth, and addition of human recombinant IL-8 rescues this antitumor effects. Our results imply the role of HMGB1 in EMT through IL-8 mediation, and a potential mechanism of GC micrometastasis. Our observations suggest combination strategy of HMGB1 and IL-8 as a promising diagnostic and therapeutic target to control GC micrometastasis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
MES solution, BioUltra, Molecular Biology, 0.5 M in H2O
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
HMG-1 human, lyophilized powder, ≥90% (SDS-PAGE), Histidine-tagged, recombinant, expressed in E. coli
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)