Merck
  • Targeting delivery of tocopherol and doxorubicin grafted-chitosan polymeric micelles for cancer therapy: In vitro and in vivo evaluation.
Targeting delivery of tocopherol and doxorubicin grafted-chitosan polymeric micelles for cancer therapy: In vitro and in vivo evaluation.
Colloids and surfaces. B, Biointerfaces (2015-06-29)
Joung-Pyo Nam, Kyeong-Jae Lee, Joung-Woo Choi, Chae-Ok Yun, Jae-Woon Nah
ABSTRAKT
In this study, we report the development of a novel, redox-sensitive chitosan-based targeted drug delivery system, containing two drugs. We determined whether the synthesized polymeric micelles (HPTOC-DOX) were suitable as a drug carrier. The formation of HPTOC-DOX micelles was confirmed by (1)H NMR. HPTOC-DOX formed micelles of approximately 151.9-311.2nm in size in aqueous solution. Analysis of the drug release profile of HPTOC-DOX in different pH conditions (pH 5.2, 6.2, and 7.4) indicated that DOX was released from HPTOC-DOX micelles at acidic pH (5.2 or 6.2), while almost no DOX was released at pH 7.4. In vitro cell cytotoxicity and hemolysis assays indicated that HPTOC-DOX micelles safely deliver anti-cancer drugs and decrease the cytotoxicity of DOX. In vitro anti-cancer activity assays, confocal laser scanning microscopy analysis of SK-BR-3 cells, and in vivo anti-tumor activity in SK-BR-3-derived tumor-bearing mice were used to evaluate synergistic drug effects and the effect of the targeting peptide (anti-human epidermal growth factor receptor 2 [HER2] target peptide, epitope form; LTVSPWY) on receptor-mediated endocytosis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
3,3′-Dithiodipropionic acid di(N-hydroxysuccinimide ester), powder
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Deuterium oxide, 60 atom % D
Sigma-Aldrich
Deuterium oxide, 70 atom % D
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, BioXtra
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, commercial grade, powder
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, ≥99.0% (AT)
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, crystalline