Merck
  • High Peritoneal Glucose Exposure Is Associated with Increased Incidence of Relapsing and Recurrent Bacterial Peritonitis in Patients Undergoing Peritoneal Dialysis.
High Peritoneal Glucose Exposure Is Associated with Increased Incidence of Relapsing and Recurrent Bacterial Peritonitis in Patients Undergoing Peritoneal Dialysis.
Blood purification (2015-07-04)
Na Jiang, Zhen Zhang, Wei Fang, Jiaqi Qian, Shan Mou, Zhaohui Ni
ABSTRAKT
We investigated the association of peritoneal glucose exposure and dialysis exchange number with peritonitis outcome in patients undergoing continuous ambulatory peritoneal dialysis (CAPD). The first episodes of bacterial peritonitis were retrospectively analyzed in 187 CAPD patients. Peritoneal glucose exposure was calculated based on PD prescription at the onset of peritonitis. Patients with peritoneal glucose exposure ≤140 g/day showed a higher and complete cure rate of peritonitis (66 vs. 51.7%, p = 0.047), lower occurrence of relapsing/recurrent peritonitis (10.0 vs. 21.8%, p = 0.026) and catheter removal (14.0 vs. 26.4%, p = 0.033). Patients who exchanged more than three times every day demonstrated marginally higher catheter removal rate (24.1 vs. 13.0%, p = 0.085). Logistic analysis indicated that peritoneal glucose exposure >140 g/day was an independent predictor for relapsing/recurrent peritonitis (RR: 1.959, p = 0.042). High peritoneal glucose exposure is associated with increased incidence of relapsing/recurrent peritonitis in CAPD patients.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)