Merck
Chitosan hydrogels enriched with polyphenols: Antibacterial activity, cell adhesion and growth and mineralization.
Carbohydrate polymers (2015-06-09)
Jana Lišková, Timothy E L Douglas, Jana Beranová, Agata Skwarczyńska, Mojca Božič, Sangram Keshari Samal, Zofia Modrzejewska, Selestina Gorgieva, Vanja Kokol, Lucie Bačáková
ABSTRAKT
Injectable hydrogels for bone regeneration consisting of chitosan, sodium beta-glycerophosphate (Na-β-GP) and alkaline phosphatase (ALP) were enriched with the polyphenols phloroglucinol (PG) and gallic acid (GA) and characterized physicochemically and biologically with respect to properties relevant for applications in bone regeneration, namely gelation kinetics, mineralizability, antioxidant properties, antibacterial activity, cytocompatibility and ability to support adhesion and growth of human osteoblast-like MG63 cells. Enrichment with PG and GA had no negative effect on gelation kinetics and mineralizability. PG and GA both enhanced antioxidant activity of unmineralized hydrogels. Mineralization reduced antioxidant activity of hydrogels containing GA. Hydrogels containing GA, PG and without polyphenols reduced colony forming ability of Escherichia coli after 1h, 3h and 6h incubation and slowed E. coli growth in liquid culture for 150min. Hydrogels containing GA were cytotoxic and supported cell growth more poorly than polyphenol-free hydrogels. PG had no negative effect on cell adhesion and growth.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Sigma-Aldrich
Diethyl azodicarboxylate solution, purum, ~40% in toluene (H-NMR)
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
1,3-Propanediol, 98%
Sigma-Aldrich
Chitosan from shrimp shells, low-viscous