Merck
Myristoylation confers noncanonical AMPK functions in autophagy selectivity and mitochondrial surveillance.
Nature communications (2015-08-15)
Jiyong Liang, Zhi-Xiang Xu, Zhiyong Ding, Yiling Lu, Qinghua Yu, Kaitlin D Werle, Ge Zhou, Yun-Yong Park, Guang Peng, Michael J Gambello, Gordon B Mills
ABSTRAKT
AMP-activated protein kinase (AMPK) plays a central role in cellular energy sensing and bioenergetics. However, the role of AMPK in surveillance of mitochondrial damage and induction of mitophagy remains unclear. We demonstrate herein that AMPK is required for efficient mitophagy. Mitochondrial damage induces a physical association of AMPK with ATG16-ATG5-12 and an AMPK-dependent recruitment of the VPS34 and ATG16 complexes with the mitochondria. Targeting AMPK to the mitochondria is both sufficient to induce mitophagy and to promote cell survival. Recruitment of AMPK to the mitochondria requires N-myristoylation of AMPKβ by the type-I N-myristoyltransferase 1 (NMT1). Our data support a spatiotemporal model wherein recruitment of AMPK in association with components of the VPS34 and ATG16 complex to damaged mitochondria regulates selective mitophagy to maintain cancer cell viability.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Tannic acid, Source: Chinese natural gall nuts
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
Tannic acid, ACS reagent
Sigma-Aldrich
Tannic acid
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Golgicide A, ≥98% (HPLC)
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 2% in H2O
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O
Sigma-Aldrich
Przeciwciało przeciw ubikwitynie, klon P4D1-A11, clone P4D1-A11, Upstate®, from mouse