Merck
  • Metal-organic framework UiO-66 modified magnetite@silica core-shell magnetic microspheres for magnetic solid-phase extraction of domoic acid from shellfish samples.
Metal-organic framework UiO-66 modified magnetite@silica core-shell magnetic microspheres for magnetic solid-phase extraction of domoic acid from shellfish samples.
Journal of chromatography. A (2015-05-23)
Wenmin Zhang, Zhiming Yan, Jia Gao, Ping Tong, Wei Liu, Lan Zhang
ABSTRAKT
Fe3O4@SiO2@UiO-66 core-shell magnetic microspheres were synthesized and characterized by transmission electron microscopy, scanning electron microscopy, X-ray diffraction, Fourier transform infrared spectrometry, vibrating sample magnetometry, nitrogen adsorption porosimetry and zeta potential analyzer. The synthesized Fe3O4@SiO2@UiO-66 microspheres were first used for magnetic solid-phase extraction (MSPE) of domoic acid (DA) in shellfish samples. Combined with high performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS), a fast, simple and sensitive method for the determination of DA was established successfully. Under the optimized conditions, the developed method showed short analysis time, good linearity (r(2) = 0.9990), low limit of detection (1.45 pg mL(-1); S/N = 3:1), low limit of quantification (4.82 pg mL(-1); S/N = 10:1), and good extraction repeatability (RSD ≤ 5.0%; n = 5). Real shellfish samples were processed using the developed method, and trace level of DA was detected. The results demonstrate that Fe3O4@SiO2@UiO-66 core-shell magnetic microspheres are the promising sorbents for rapid and efficient extraction of polar analytes from complex biological samples.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Sodium acetate, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Methyl Orange, ACS reagent, Dye content 85 %
Sigma-Aldrich
Terephthalic acid, 98%
Sigma-Aldrich
Sodium acetate solution, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glutaric anhydride, 95%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)