Merck
Down-regulated CFTR During Aging Contributes to Benign Prostatic Hyperplasia.
Journal of cellular physiology (2014-12-30)
Chen Xie, Xiao Sun, Jing Chen, Chi Fai Ng, Kin Mang Lau, Zhiming Cai, Xiaohua Jiang, Hsiao Chang Chan
ABSTRAKT
Benign prostatic hyperplasia (BPH) is a hyper-proliferative disease of the aging prostate; however, the exact mechanism underlying the development of BPH remains incompletely understood. The present study investigated the possible involvement of the cystic fibrosis transmembrane conductance regulator (CFTR), which has been previously shown to negatively regulate nuclear factor-κB (NF-κB)/cyclooxygenase 2 (COX2)/prostaglandin E2 (PGE2) pathway, in the pathogenesis of BPH. Our results showed decreasing CFTR and increasing COX2 expression in rat prostate tissues with aging. Furthermore, suppression of CFTR led to increased expression of COX2 and over-production of PGE2 in a normal human prostate epithelial cell line (PNT1A) with elevated NF-κB activity. PGE2 stimulated the proliferation of primary rat prostate stromal cells but not epithelial cells, with increased PCNA expression. In addition, the condition medium from PNT1A cells after inhibition or knockdown of CFTR promoted cell proliferation of prostate stromal cells which could be reversed by COX2 or NF-κB inhibitor. More importantly, the involvement of CFTR in BPH was further demonstrated by the down-regulation of CFTR and up-regulation of COX2/NF-κB in human BPH samples. The present results suggest that CFTR may be involved in regulating PGE2 production through its negative regulation on NF-κB/COX2 pathway in prostate epithelial cells, which consequently stimulates cell growth of prostate stromal cells. The overstimulation of prostate stromal cell proliferation by down-regulation of CFTR-enhanced PGE2 production and release during aging may contribute to the development of BPH.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium trichloroacetate, 97%
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
NS-398, ≥98% (HPLC), solid
Sigma-Aldrich
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
Sigma-Aldrich
DAPI, for nucleic acid staining